siRNA Details
| Gene Symbol: WDR75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303096.1 | NP_001290025.1 | 15 | 1656 | ||
| NM_032168.2 | NP_115544.1 | 14 | 1622 | |||
| GenBank mRNA | AK022581.1 | - | 14 | 1569 | ||
| AK022607.1 | - | 14 | 1581 | |||
| AK023276.1 | - | 11 | 1261 | |||
| AK091546.1 | - | 15 | 1583 | |||
| AK290445.1 | - | 14 | 1573 | |||
| AL122097.1 | - | 10 | 1189 | |||
| BC040567.1 | - | 14 | 1622 | 5267120 | ||
| CR749440.1 | - | 14 | 3231 | |||

WDR75
Genomic Map