siRNA Details
| Gene Symbol: EVA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135032.1 | NP_001128504.1 | 4 | 1816 | ||
| NM_032181.2 | NP_115557.1 | 4 | 1776 | |||
| GenBank mRNA | AK023453.1 | - | 4 | 1778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EVA1A| Gene Symbol: EVA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135032.1 | NP_001128504.1 | 4 | 1816 | ||
| NM_032181.2 | NP_115557.1 | 4 | 1776 | |||
| GenBank mRNA | AK023453.1 | - | 4 | 1778 | ||