siRNA Details
| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032288.6 | NP_115664.2 | 1 | 305 | ||
| GenBank mRNA | AJ344094.1 | - | 1 | 151 | ||
| AK027672.1 | - | 1 | 189 | |||
| AK296283.1 | - | 1 | 120 | |||
| AK315019.1 | - | 1 | 154 | |||
| AL136558.1 | - | 1 | 158 | |||
| BC035006.2 | - | 1 | 308 | 5267824 | ||
| BC039734.1 | - | 1 | 215 | 6042926 | ||
| BX509179.1 | - | 1 | 155 | |||
| CR533502.1 | - | 1 | 86 | |||

FYTTD1
Genomic Map