siRNA Details
| Gene Symbol: LRCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032773.3 | NP_116162.1 | 3 | 475 | ||
| XM_005269362.2 | XP_005269419.1 | 3 | 846 | |||
| XM_005269363.2 | XP_005269420.1 | 3 | 846 | |||
| XM_005269365.2 | XP_005269422.1 | 3 | 846 | |||
| XM_005269367.2 | XP_005269424.1 | 3 | 846 | |||
| XM_006713791.3 | XP_006713854.1 | 3 | 846 | |||
| XM_006713792.3 | XP_006713855.1 | 3 | 846 | |||
| XM_006713793.2 | XP_006713856.1 | 3 | 846 | |||
| XM_011513242.2 | XP_011511544.1 | 3 | 846 | |||
| XM_011513243.2 | XP_011511545.1 | 3 | 846 | |||
| XM_017007350.1 | XP_016862839.1 | 3 | 846 | |||
| XM_017007351.1 | XP_016862840.1 | 3 | 846 | |||
| XM_017007352.1 | XP_016862841.1 | 3 | 846 | |||
| GenBank mRNA | AK303521.1 | - | 3 | 437 | ||
| BC007504.2 | - | 3 | 427 | 2960711 | ||

LRCH3
Genomic Map