siRNA Details
| Gene Symbol: KLHL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032775.3 | NP_116164.2 | 3 | 518 | ||
| XM_017029019.1 | XP_016884508.1 | 3 | 814 | |||
| XM_017029020.1 | XP_016884509.1 | 4 | 1993 | |||
| XM_017029021.1 | XP_016884510.1 | 3 | 447 | |||
| XM_017029025.1 | XP_016884514.1 | 2 | 166 | |||
| GenBank mRNA | AK027266.1 | - | 3 | 457 | ||
| AK222850.1 | - | 3 | 424 | |||
| AK290669.1 | - | 3 | 430 | |||
| BC015923.1 | - | 3 | 470 | 4562420 | ||
| CR456352.1 | - | 2 | 395 | |||
| DA381792.1 | - | 3 | 518 | |||

KLHL22
Genomic Map