siRNA Details
| Gene Symbol: FAM136A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032822.2 | NP_116211.2 | 3 | 1070 | ||
| XM_005264620.1 | XP_005264677.1 | 4 | 1120 | |||
| GenBank mRNA | AK027574.1 | - | 3 | 1050 | ||
| AK225151.1 | - | 3 | 1038 | |||
| AL833147.1 | - | 3 | 1013 | |||

FAM136A
Genomic Map