siRNA Details
| Gene Symbol: TMEM87B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032824.2 | NP_116213.1 | 1, 2 | 539 | ||
| XM_005263827.2 | XP_005263884.1 | 1, 2 | 496 | |||
| XM_006712807.2 | XP_006712870.1 | 1, 2 | 496 | |||
| XM_017005121.1 | XP_016860610.1 | 1, 2 | 42 | |||
| GenBank mRNA | AK027587.1 | - | 1, 2 | 524 | ||
| AK290294.1 | - | 1, 2 | 185 | |||
| AY429534.1 | - | 1, 2 | 42 | |||
| AY429535.1 | - | 1, 2 | 42 | |||
| BC115373.1 | - | 1, 2 | 343 | 40026807 | ||
| DA806277.1 | - | 1, 2 | 539 | |||

TMEM87B
Genomic Map