siRNA Details
| Gene Symbol: MPND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159846.2 | NP_001153318.1 | 10 | 1325 | ||
| NM_001300862.1 | NP_001287791.1 | 12 | 1475 | |||
| NM_032868.5 | NP_116257.2 | 11 | 1385 | |||
| XM_006722926.2 | XP_006722989.1 | 12 | 1349 | |||
| GenBank mRNA | AK027887.1 | - | 10 | 1298 | ||
| BC032652.1 | - | 11 | 1342 | 5503607 | ||
| CR977937.1 | - | 9 | 814 | |||

MPND
Genomic Map