siRNA Details
| Gene Symbol: TMEM128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297551.1 | NP_001284480.1 | 5 | 995 | ||
| NM_001297552.1 | NP_001284481.1 | 5 | 993 | |||
| NM_032927.3 | NP_116316.1 | 5 | 1500 | |||
| XM_005248034.2 | XP_005248091.1 | 5 | 1839 | |||
| GenBank mRNA | AK001453.1 | - | 1 | 645 | ||
| BC007729.2 | - | 5 | 1465 | 4303698 | ||
| BC091482.1 | - | 5 | 995 | 30344390 | ||
| CR933611.1 | - | 3 | 4939 | |||

TMEM128
Genomic Map