siRNA Details
| Gene Symbol: HELZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037335.2 | NP_001032412.2 | 17 | 8352 | ||
| NM_033405.3 | NP_208384.3 | 11 | 5857 | |||
| GenBank mRNA | AB051556.2 | - | 11 | 5857 | ||
| AB201715.1 | - | 16 | 7530 | |||
| AF517673.1 | - | 11 | 6177 | |||
| AK055611.1 | - | 6 | 889 | |||
| AK074171.1 | - | 9 | 2886 | |||
| BC036883.1 | - | 9 | 1436 | |||
| BC150283.1 | - | 11 | 5857 | 9007272 | ||

HELZ2
Genomic Map