siRNA Details
| Gene Symbol: FDX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031734.3 | NP_001026904.2 | 5 | 594 | ||
| GenBank mRNA | AK300568.1 | - | 5 | 587 | ||
| BC063460.1 | - | 5 | 578 | 5179971 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FDX1L| Gene Symbol: FDX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031734.3 | NP_001026904.2 | 5 | 594 | ||
| GenBank mRNA | AK300568.1 | - | 5 | 587 | ||
| BC063460.1 | - | 5 | 578 | 5179971 | ||