siRNA Details
| Gene Symbol: CFAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282761.1 | NP_001269690.1 | 3 | 427 | ||
| NM_080667.6 | NP_542398.3 | 2 | 352 | |||
| XM_017003254.1 | XP_016858743.1 | 2 | 343 | |||
| XM_017003255.1 | XP_016858744.1 | 2 | 366 | |||
| GenBank mRNA | AK309764.1 | - | 2 | 301 | ||
| AY358097.1 | - | 3 | 419 | |||
| BC001797.1 | - | 282 | ||||
| BC010011.2 | - | 2 | 259 | 4309613 | ||
| DB448836.1 | - | 2 | 351 | |||

CFAP36
Genomic Map