siRNA Details
| Gene Symbol: DCBLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080927.3 | NP_563615.3 | 2 | 619 | ||
| GenBank mRNA | AB073146.1 | - | 2 | 619 | ||
| AF387547.1 | - | 2 | 267 | |||
| AK056350.1 | - | 2 | 612 | |||
| AK302649.1 | - | 2 | 99 | |||
| BC007117.1 | - | 3 | 166 | |||
| BC029658.1 | - | 2 | 138 | 4431275 | ||
| D29810.1 | - | 1 | 51 | |||

DCBLD2
Genomic Map