siRNA Details
| Gene Symbol: FAM114A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138389.2 | NP_612398.2 | 3 | 522 | ||
| XM_005262672.2 | XP_005262729.1 | 2 | 328 | |||
| XM_017008838.1 | XP_016864327.1 | 2 | 328 | |||
| GenBank mRNA | AK292831.1 | - | 3 | 449 | ||
| BC001096.2 | - | 1 | 273 | 3507281 | ||
| BC040452.1 | - | 3 | 439 | 3846822 | ||
| DA802033.1 | - | 2 | 373 | |||

FAM114A1
Genomic Map