siRNA Details
| Gene Symbol: TMEM169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142310.1 | NP_001135782.1 | 4 | 3079 | ||
| NM_001142311.1 | NP_001135783.1 | 3 | 2993 | |||
| NM_001142312.1 | NP_001135784.1 | 3 | 2987 | |||
| NM_138390.3 | NP_612399.1 | 3 | 2989 | |||
| XM_006712851.1 | XP_006712914.1 | 3 | 3193 | |||
| XM_011512173.2 | XP_011510475.1 | 3 | 3384 | |||
| GenBank mRNA | AK091582.1 | - | 3 | 2936 | ||
| AK172731.1 | - | 1 | 2149 | |||

TMEM169
Genomic Map