siRNA Details
Gene Symbol: SHKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138392.3 | NP_612401.2 | 3, 4 | 241 | ||
XM_006723474.1 | XP_006723537.1 | 3, 4 | 241 | |||
XM_006723475.1 | XP_006723538.1 | 3, 4 | 241 | |||
XM_011527492.1 | XP_011525794.1 | 3, 4 | 241 | |||
XM_011527493.1 | XP_011525795.1 | 2, 3 | 106 | |||
XM_017027473.1 | XP_016882962.1 | 3, 4 | 241 | |||
XM_017027474.1 | XP_016882963.1 | 2, 3 | 106 | |||
GenBank mRNA | AF258553.1 | - | 3, 4 | 240 | ||
AK075057.1 | - | 3, 4 | 237 | |||
AK123695.1 | - | 1, 2 | 219 | |||
AK300808.1 | - | 2, 3 | 179 | |||
AK300819.1 | - | 3, 4 | 237 | |||
AK312748.1 | - | 3, 4 | 205 | |||
BC022855.1 | - | 3, 4 | 212 | 4102901 | ||
DB038927.1 | - | 3, 4 | 241 |
