siRNA Details
| Gene Symbol: TMIGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308232.1 | NP_001295161.1 | 3 | 231 | ||
| NM_144615.2 | NP_653216.2 | 4 | 591 | |||
| XM_017026284.1 | XP_016881773.1 | 4 | 599 | |||
| GenBank mRNA | BC015655.1 | - | 4 | 591 | 4811204 | |
| HQ452865.1 | - | 3 | 185 | |||

TMIGD2
Genomic Map