siRNA Details
| Gene Symbol: TMIGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169126.1 | NP_001162597.1 | 5 | 922 | ||
| NM_001308232.1 | NP_001295161.1 | 4 | 574 | |||
| NM_144615.2 | NP_653216.2 | 5 | 934 | |||
| GenBank mRNA | AY358964.1 | - | 5 | 884 | ||
| BC015655.1 | - | 5 | 934 | 4811204 | ||

TMIGD2
Genomic Map