siRNA Details
| Gene Symbol: TMEM42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144638.2 | NP_653239.1 | 2 | 221 | ||
| GenBank mRNA | AL532443.3 | - | 3 | 212 | ||
| AL834253.1 | - | 1 | 2306 | |||
| BC019851.1 | - | 2 | 214 | 5114515 | ||
| DB499198.1 | - | 2 | 221 | |||

TMEM42
Genomic Map