siRNA Details
| Gene Symbol: SNRPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003090.3 | NP_003081.2 | 6 | 580 | ||
| GenBank mRNA | AK090986.1 | - | 2 | 1528 | ||
| AK223070.1 | - | 6 | 537 | |||
| AK223110.1 | - | 6 | 502 | |||
| AK290078.1 | - | 5 | 1150 | |||
| AK312200.1 | - | 6 | 511 | |||
| BC022816.1 | - | 6 | 533 | 4904205 | ||
| BC067846.1 | - | 5 | 1163 | 30330402 | ||
| BC071717.1 | - | 1 | 488 | 4514975 | ||
| X13482.1 | - | 6 | 532 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021336, BC013777, AK206837, BC029642, AK153502, and others.

SNRPA1
Genomic Map