siRNA Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122870.2 | NP_001116342.1 | 6 | 599 | ||
| NM_144641.3 | NP_653242.3 | 7 | 965 | |||
| XM_005264879.2 | XP_005264936.1 | 7 | 1033 | |||
| XM_005264880.3 | XP_005264937.1 | 6 | 778 | |||
| XM_005264881.1 | XP_005264938.1 | 7 | 854 | |||
| GenBank mRNA | AB474372.1 | - | 7 | 933 | ||
| AK056894.1 | - | 7 | 854 | |||
| AK129647.1 | - | 1 | 230 | |||
| AL832484.1 | - | 5 | 873 | |||
| BC009644.1 | - | 6 | 560 | 3905720 | ||

PPM1M
Genomic Map