siRNA Details
| Gene Symbol: SPICE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144718.3 | NP_653319.1 | 7 | 822 | ||
| XM_011512461.2 | XP_011510763.1 | 7 | 579 | |||
| XM_017005775.1 | XP_016861264.1 | 7 | 579 | |||
| XM_017005776.1 | XP_016861265.1 | 7 | 716 | |||
| XM_017005777.1 | XP_016861266.1 | 7 | 639 | |||
| XM_017005778.1 | XP_016861267.1 | 7 | 823 | |||
| GenBank mRNA | AK126897.1 | - | 3 | 228 | ||
| AL832148.1 | - | 7 | 791 | |||
| AL832327.2 | - | 7 | 605 | |||
| AY099107.1 | - | 7 | 742 | |||
| BC036951.1 | - | 7 | 763 | 5500345 | ||

SPICE1
Genomic Map