siRNA Details
| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144736.4 | NP_653337.1 | 2, 3 | 294 | ||
| XM_005264410.1 | XP_005264467.1 | 2, 3 | 274 | |||
| XM_005264411.1 | XP_005264468.1 | 2, 3 | 272 | |||
| XM_011532956.1 | XP_011531258.1 | 2, 3 | 262 | |||
| GenBank mRNA | AK294240.1 | - | 2, 3 | 228 | ||
| AK299015.1 | - | 2, 3 | 268 | |||
| AK303555.1 | - | 2, 3 | 274 | |||
| BC004548.2 | - | 2, 3 | 259 | 3959139 | ||
| BI334629.1 | - | 2, 3 | 245 | |||
| BM172474.1 | - | 2, 3 | 299 | |||

NDUFAF7
Genomic Map