siRNA Details
| Gene Symbol: CCDC151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302453.1 | NP_001289382.1 | 12 | 1470 | ||
| NM_001302454.1 | NP_001289383.1 | 10 | 1584 | |||
| NM_145045.4 | NP_659482.3 | 12 | 1764 | |||
| GenBank mRNA | AK056363.1 | - | 12 | 1685 | ||
| AK294355.1 | - | 14 | 1664 | |||
| AK302113.1 | - | 12 | 1471 | |||
| AK308072.1 | - | 10 | 1546 | |||
| BC014252.2 | - | 11 | 1321 | |||
| BC141828.1 | - | 12 | 1696 | 40148425 | ||
| BC142637.1 | - | 12 | 1709 | 40147788 | ||
| BC144421.1 | - | 10 | 1243 | |||

CCDC151
Genomic Map