siRNA Details
| Gene Symbol: EVI5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159944.2 | NP_001153416.1 | 9, 10 | 1182 | ||
| NM_145245.4 | NP_660288.1 | 10, 11 | 1293 | |||
| XM_005272458.4 | XP_005272515.1 | 10, 11 | 1287 | |||
| XM_017026239.1 | XP_016881728.1 | 10, 11 | 1279 | |||
| GenBank mRNA | AB449874.1 | - | 9, 10 | 1139 | ||
| AK091609.1 | - | 4, 5 | 331 | |||
| BC014111.2 | - | 10, 11 | 1211 | 4542709 | ||

EVI5L
Genomic Map