siRNA Details
| Gene Symbol: FAAP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300978.1 | NP_001287907.1 | 3 | 450 | ||
| NM_152266.4 | NP_689479.1 | 5 | 706 | |||
| XM_005259393.3 | XP_005259450.1 | 5 | 653 | |||
| GenBank mRNA | AK128668.1 | - | 5 | 673 | ||
| BC003535.1 | - | 5 | 626 | 3609326 | ||
| BC010170.2 | - | 5 | 634 | 3659284 | ||
| BC020247.1 | - | 5 | 654 | 4620233 | ||
| CN368337.1 | - | 3 | 368 | |||
| KU178830.1 | - | 4 | 563 | |||
| KU178831.1 | - | 4 | 505 | |||

FAAP24
Genomic Map