siRNA Details
| Gene Symbol: TTC30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152275.3 | NP_689488.3 | 1 | 2609 | ||
| GenBank mRNA | AK057530.1 | - | 2 | 2246 | ||
| AK125104.1 | - | 2 | 2313 | |||
| AL049329.1 | - | 1 | 709 | |||
| BC042848.1 | - | 1 | 2613 | 4831326 | ||
| BC048006.1 | - | 1 | 2383 | 5299020 | ||

TTC30A
Genomic Map