siRNA Details
| Gene Symbol: IZUMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321449.1 | NP_001308378.1 | 2 | 368 | ||
| NM_152358.2 | NP_689571.2 | 2 | 368 | |||
| XM_011526436.2 | XP_011524738.1 | 2 | 308 | |||
| XM_011526437.2 | XP_011524739.1 | 2 | 323 | |||
| GenBank mRNA | AK131024.1 | - | 2 | 368 | ||
| AK307321.1 | - | 2 | 214 | |||
| AY358196.1 | - | 2 | 282 | |||
| BC029535.1 | - | 2 | 249 | |||
| DB523757.2 | - | 2 | 140 | |||

IZUMO2
Genomic Map