siRNA Details
| Gene Symbol: ERICH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308234.1 | NP_001295163.1 | 6 | 507 | ||
| NM_152394.4 | NP_689607.2 | 6 | 811 | |||
| XM_005247120.3 | XP_005247177.1 | 6 | 1403 | |||
| GenBank mRNA | AK093375.1 | - | 7 | 646 | ||
| BC029577.1 | - | 6 | 491 | 5268183 | ||
| BC036239.1 | - | 6 | 795 | 5297417 | ||

ERICH6
Genomic Map