siRNA Details
| Gene Symbol: IQCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152397.2 | NP_689610.2 | 2 | 180 | ||
| GenBank mRNA | AK131018.1 | - | 2 | 131 | ||
| BC029595.1 | - | 2 | 145 | 5273825 | ||
| BC034228.1 | - | 2 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IQCF1| Gene Symbol: IQCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152397.2 | NP_689610.2 | 2 | 180 | ||
| GenBank mRNA | AK131018.1 | - | 2 | 131 | ||
| BC029595.1 | - | 2 | 145 | 5273825 | ||
| BC034228.1 | - | 2 | 181 | |||