siRNA Details
| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 5 | 1595 | |||
| NR_110321.1 | 5 | 1305 | ||||
| GenBank mRNA | AK058057.1 | - | 5 | 1553 | ||
| BC031284.1 | - | 5 | 1301 | |||
| BC069223.1 | - | 5 | 1526 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00905| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 5 | 1595 | |||
| NR_110321.1 | 5 | 1305 | ||||
| GenBank mRNA | AK058057.1 | - | 5 | 1553 | ||
| BC031284.1 | - | 5 | 1301 | |||
| BC069223.1 | - | 5 | 1526 | |||