siRNA Details
| Gene Symbol: HORMAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529917.2 | XP_011528219.1 | 11 | 1606 | ||
| GenBank mRNA | BC030013.1 | - | 11 | 1357 | 4823215 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HORMAD2| Gene Symbol: HORMAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529917.2 | XP_011528219.1 | 11 | 1606 | ||
| GenBank mRNA | BC030013.1 | - | 11 | 1357 | 4823215 | |