siRNA Details
| Gene Symbol: ENTHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152512.3 | NP_689725.2 | 7 | 1917 | ||
| XM_006724149.3 | XP_006724212.1 | 7 | 2512 | |||
| XM_006724150.3 | XP_006724213.1 | 7 | 1702 | |||
| XM_006724151.1 | XP_006724214.1 | 6 | 1406 | |||
| XM_011529927.2 | XP_011528229.1 | 7 | 1948 | |||
| XM_011529928.2 | XP_011528230.1 | 6 | 1768 | |||
| XM_011529930.2 | XP_011528232.1 | 3 | 933 | |||
| GenBank mRNA | AK058150.1 | - | 2 | 599 | ||
| AK093154.1 | - | 5 | 1151 | |||
| BC033895.2 | - | 7 | 1917 | 4831324 | ||
| CR456452.1 | - | 6 | 1712 | |||

ENTHD1
Genomic Map