siRNA Details
| Gene Symbol: LINC01106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027244.1 | 2 | 1136 | |||
| GenBank mRNA | AK095678.1 | - | 2 | 1446 | ||
| CR749351.1 | - | 3 | 1229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01106| Gene Symbol: LINC01106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027244.1 | 2 | 1136 | |||
| GenBank mRNA | AK095678.1 | - | 2 | 1446 | ||
| CR749351.1 | - | 3 | 1229 | |||