siRNA Details
| Gene Symbol: C3orf30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152539.2 | NP_689752.2 | 1 | 394 | ||
| GenBank mRNA | AK057421.1 | - | 1 | 394 | ||
| BC130475.1 | - | 1 | 393 | 40146506 | ||
| BC144197.1 | - | 1 | 393 | 9052717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3orf30| Gene Symbol: C3orf30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152539.2 | NP_689752.2 | 1 | 394 | ||
| GenBank mRNA | AK057421.1 | - | 1 | 394 | ||
| BC130475.1 | - | 1 | 393 | 40146506 | ||
| BC144197.1 | - | 1 | 393 | 9052717 | ||