siRNA Details
| Gene Symbol: AGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018238.3 | NP_060708.1 | 4 | 365 | ||
| XM_005250023.3 | XP_005250080.1 | 4 | 261 | |||
| XM_011516397.2 | XP_011514699.1 | 4 | 831 | |||
| GenBank mRNA | AJ278150.1 | - | 3 | 221 | ||
| AK001704.1 | - | 4 | 233 | |||
| AK222769.1 | - | 4 | 227 | |||
| AK299131.1 | - | 4 | 233 | |||
| AK304502.1 | - | 5 | 252 | |||
| BC022777.1 | - | 3 | 219 | 3913448 | ||
| DB032353.1 | - | 4 | 365 | |||

AGK
Genomic Map