siRNA Details
| Gene Symbol: PI4K2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018323.3 | NP_060793.2 | 2, 3 | 552 | ||
| XM_005248174.1 | XP_005248231.1 | 2, 3 | 550 | |||
| XM_005248175.4 | XP_005248232.1 | 2, 3 | 330 | |||
| GenBank mRNA | AF411320.1 | - | 2, 3 | 419 | ||
| AK001967.1 | - | 2, 3 | 540 | |||
| AK128622.1 | - | 3, 4 | 457 | |||
| AY065990.1 | - | 2, 3 | 530 | |||
| AY091514.1 | - | 2, 3 | 539 | |||
| BC051749.1 | - | 2, 3 | 552 | 6050442 | ||

PI4K2B
Genomic Map