siRNA Details
| Gene Symbol: PLCXD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185106.1 | NP_001172035.1 | 3 | 1288 | ||
| NM_153268.3 | NP_695000.1 | 3 | 1288 | |||
| GenBank mRNA | AK056141.1 | - | 3 | 1288 | ||
| BC121155.1 | - | 3 | 1042 | 40121377 | ||
| BC121156.1 | - | 3 | 1042 | 40121378 | ||

PLCXD2
Genomic Map