siRNA Details
| Gene Symbol: RGL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153615.1 | NP_705843.1 | 5 | 2149 | ||
| GenBank mRNA | AK131340.1 | - | 7 | 2587 | ||
| AY101396.1 | - | 5 | 2149 | |||
| BC101108.3 | - | 4 | 1829 | 40011557 | ||
| BC101109.2 | - | 4 | 1829 | 40011564 | ||
| BC101110.3 | - | 5 | 1704 | |||

RGL4
Genomic Map