siRNA Details
| Gene Symbol: LINC00471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024079.1 | 3 | 533 | |||
| GenBank mRNA | BC033054.2 | - | 3 | 533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00471| Gene Symbol: LINC00471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024079.1 | 3 | 533 | |||
| GenBank mRNA | BC033054.2 | - | 3 | 533 | ||