siRNA Details
| Gene Symbol: ZNF776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173632.3 | NP_775903.3 | 3 | 4561 | ||
| GenBank mRNA | AK127764.1 | - | 2 | 4205 | ||
| BX957217.1 | - | 3 | 4514 | |||
| CR749390.1 | - | 3 | 4514 | |||
| CR936802.1 | - | 3 | 4515 | |||

ZNF776
Genomic Map