siRNA Details
| Gene Symbol: C20orf197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302813.1 | NP_001289742.1 | 3 | 1762 | ||
| NM_001302815.1 | NP_001289744.1 | 4 | 1451 | |||
| NM_173644.2 | NP_775915.1 | 4 | 1842 | |||
| XM_017027828.1 | XP_016883317.1 | 3 | 10567 | |||
| XM_017027829.1 | XP_016883318.1 | 4 | 8229 | |||
| XM_017027830.1 | XP_016883319.1 | 3 | 8094 | |||
| XM_017027831.1 | XP_016883320.1 | 4 | 9913 | |||
| XM_017027832.1 | XP_016883321.1 | 4 | 1966 | |||
| GenBank mRNA | AK091179.1 | - | 4 | 1842 | ||
| AY358539.1 | - | 1 | 3551 | |||
| BC101349.1 | - | 3 | 1708 | 40025393 | ||
| BC101350.1 | - | 3 | 1317 | 40025394 | ||
| BC101351.2 | - | 3 | 1708 | 40025395 | ||

C20orf197
Genomic Map