siRNA Details
| Gene Symbol: DNAH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001370.1 | NP_001361.1 | 11 | 1839 | ||
| XM_006711956.2 | XP_006712019.1 | 11 | 1944 | |||
| XM_011532649.2 | XP_011530951.1 | 11 | 1793 | |||
| XM_011532650.2 | XP_011530952.1 | 11 | 1794 | |||
| XM_011532652.1 | XP_011530954.1 | 11 | 1839 | |||
| XM_011532653.1 | XP_011530955.1 | 11 | 1839 | |||
| XM_011532654.1 | XP_011530956.1 | 10 | 1572 | |||
| XM_011532655.2 | XP_011530957.1 | 7 | 861 | |||
| XM_011532657.1 | XP_011530959.1 | 5 | 600 | |||
| XM_011532660.1 | XP_011530962.1 | 11 | 1839 | |||
| XM_017003521.1 | XP_016859010.1 | 11 | 1839 | |||
| XM_017003522.1 | XP_016859011.1 | 6 | 1665 | |||
| XM_017003523.1 | XP_016859012.1 | 6 | 832 | |||
| XM_017003524.1 | XP_016859013.1 | 11 | 1839 | |||
| GenBank mRNA | AK094676.1 | - | 7 | 1052 | ||
| BC104884.1 | - | 5 | 745 | |||
| BC113424.1 | - | 5 | 745 | |||
| BC143666.1 | - | 5 | 745 | |||

DNAH6
Genomic Map