siRNA Details
| Gene Symbol: SH3BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122681.1 | NP_001116153.1 | 13 | 3049 | ||
| NM_001145855.1 | NP_001139327.1 | 13 | 3192 | |||
| NM_001145856.1 | NP_001139328.1 | 13 | 3139 | |||
| NM_003023.4 | NP_003014.3 | 13 | 3190 | |||
| GenBank mRNA | AB000462.1 | - | 13 | 3189 | ||

SH3BP2
Genomic Map