siRNA Details
| Gene Symbol: MCEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174918.2 | NP_777578.2 | 2 | 138 | ||
| GenBank mRNA | AF461155.1 | - | 2 | 567 | ||
| AF461156.1 | - | 2 | 117 | |||
| BC035847.1 | - | 2 | 80 | |||
| BC104796.1 | - | 2 | 138 | 8143799 | ||
| BC104798.1 | - | 2 | 138 | 8143801 | ||

MCEMP1
Genomic Map