siRNA Details
Gene Symbol: MCEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174918.2 | NP_777578.2 | 7 | 714 | ||
GenBank mRNA | AF461155.1 | - | 7 | 1141 | ||
BC035847.1 | - | 7 | 655 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MCEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174918.2 | NP_777578.2 | 7 | 714 | ||
GenBank mRNA | AF461155.1 | - | 7 | 1141 | ||
BC035847.1 | - | 7 | 655 |