siRNA Details
| Gene Symbol: SMIM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317896.1 | NP_001304825.1 | 5 | 697 | ||
| NM_001317897.1 | NP_001304826.1 | 4 | 406 | |||
| NM_174921.2 | NP_777581.1 | 4 | 552 | |||
| XM_011513640.2 | XP_011511942.1 | 4 | 685 | |||
| XM_017007862.1 | XP_016863351.1 | 5 | 601 | |||
| XM_017007863.1 | XP_016863352.1 | 4 | 442 | |||
| GenBank mRNA | AK023351.1 | - | 4 | 415 | ||
| BC008502.1 | - | 4 | 415 | 4295121 | ||
| BQ049060.1 | - | 4 | 397 | |||

SMIM14
Genomic Map