siRNA Details
| Gene Symbol: SMIM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317896.1 | NP_001304825.1 | 5, 6 | 702 | ||
| NM_001317897.1 | NP_001304826.1 | 4, 5 | 411 | |||
| NM_174921.2 | NP_777581.1 | 4, 5 | 557 | |||
| XM_011513640.2 | XP_011511942.1 | 4, 5 | 690 | |||
| XM_017007862.1 | XP_016863351.1 | 5, 6 | 606 | |||
| XM_017007863.1 | XP_016863352.1 | 4, 5 | 447 | |||
| GenBank mRNA | AK023351.1 | - | 4, 5 | 420 | ||
| BC008502.1 | - | 4, 5 | 420 | 4295121 | ||
| BQ049060.1 | - | 4, 5 | 402 | |||

SMIM14
Genomic Map