siRNA Details
| Gene Symbol: LYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175735.3 | NP_783862.2 | 4 | 356 | ||
| XM_017003751.1 | XP_016859240.1 | 5 | 1521 | |||
| GenBank mRNA | AF323919.1 | - | 4 | 381 | ||
| AY026238.1 | - | 4 | 381 | |||
| BC100882.3 | - | 3 | 264 | 40003264 | ||
| BC100883.2 | - | 3 | 264 | 40003266 | ||
| BC100884.1 | - | 3 | 264 | 40003273 | ||
| BC100885.2 | - | 3 | 264 | 40003274 | ||
| BQ187749.1 | - | 5 | 410 | |||

LYG2
Genomic Map