siRNA Details
| Gene Symbol: SHISA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145176.1 | NP_001138648.1 | 4 | 5692 | ||
| GenBank mRNA | AK091660.1 | - | 1 | 1837 | ||
| AK094959.1 | - | 1 | 2312 | |||
| AK127223.1 | - | 1 | 3864 | |||
| AL831832.1 | - | 1 | 1748 | |||

SHISA7
Genomic Map